Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285021.1 | NP_001271950.1 | 7 - 8 | 660 | 97 | |
| XM_015430656.1 | XP_015286142.1 | 7 - 8 | 732 | 97 | ||
| GenBank mRNA | AB169387.1 | - | 7 - 8 | 660 | 97 | |
More Information
Set Membership: |
|

DENR
Genomic Map