Assay Details
| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285363.1 | NP_001272292.1 | 7 - 8 | 1057 | 102 | |
| GenBank mRNA | AB174572.1 | - | 7 - 8 | 1057 | 102 | |
More Information
Set Membership: |
|

RPP14
Genomic Map