Assay Details
| Gene Symbol: SERPINB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285378.1 | NP_001272307.1 | 4 - 5 | 554 | 57 | |
| GenBank mRNA | AB179304.1 | - | 4 - 5 | 554 | 57 | |
More Information
Set Membership: |
|

SERPINB6
Genomic Map