Assay Details
| Gene Symbol: TFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283513.1 | NP_001270442.1 | 5 - 6 | 753 | 72 | |
| XM_005548292.2 | XP_005548349.1 | 5 - 6 | 749 | 72 | ||
| XM_005548293.2 | XP_005548350.1 | 5 - 6 | 971 | 72 | ||
| XM_005548294.2 | XP_005548351.1 | 5 - 6 | 1018 | 72 | ||
| XM_015445320.1 | XP_015300806.1 | 5 - 6 | 752 | 72 | ||
| GenBank mRNA | AB169691.1 | - | 5 - 6 | 753 | 72 | |
| AB171066.1 | - | 5 - 6 | 691 | 72 | ||
More Information
Set Membership: |
|

TFG
Genomic Map