Assay Details
| Gene Symbol: DDX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284832.1 | NP_001271761.1 | 1 | 1523 | 102 | |
| GenBank mRNA | AB169808.1 | - | 1 | 1523 | 102 | |
More Information
Set Membership: |
|

DDX28
Genomic Map