Assay Details
Gene Symbol: NKAIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284846.1 | NP_001271775.1 | 4 - 5 | 578 | 69 | |
XM_015454769.1 | XP_015310255.1 | 2 - 3 | 5149 | 69 | ||
XM_015454770.1 | XP_015310256.1 | 2 - 3 | 5152 | 69 | ||
GenBank mRNA | AB174345.1 | - | 4 - 5 | 579 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
