Assay Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283245.1 | NP_001270174.1 | 1 - 2 | 129 | 103 | |
| XM_005550947.2 | XP_005551004.1 | 1 - 2 | 181 | 103 | ||
| GenBank mRNA | AB171884.1 | - | 1 - 2 | 129 | 103 | |
More Information
Set Membership: |
|

NDUFB2
Genomic Map