Assay Details
| Gene Symbol: GK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283558.1 | NP_001270487.1 | 1 | 1421 | 75 | |
| GenBank mRNA | AB178979.1 | - | 1 | 1421 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GK2| Gene Symbol: GK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283558.1 | NP_001270487.1 | 1 | 1421 | 75 | |
| GenBank mRNA | AB178979.1 | - | 1 | 1421 | 75 | |
Set Membership: |
|