Assay Details
| Gene Symbol: MCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283850.1 | NP_001270779.1 | 1 - 2 | 779 | 90 | |
| GenBank mRNA | AB172745.1 | - | 1 - 2 | 779 | 90 | |
More Information
Set Membership: |
|

MCL1
Genomic Map