Assay Details
| Gene Symbol: NAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283851.1 | NP_001270780.1 | 9 - 10 | 901 | 104 | |
| XM_015430663.1 | XP_015286149.1 | 8 - 9 | 104 | |||
| XM_015430664.1 | XP_015286150.1 | 9 - 10 | 104 | |||
| XM_015430665.1 | XP_015286151.1 | 9 - 10 | 104 | |||
| XM_015430666.1 | XP_015286152.1 | 9 - 10 | 104 | |||
| XM_015430667.1 | XP_015286153.1 | 9 - 10 | 104 | |||
| XM_015430668.1 | XP_015286154.1 | 7 - 8 | 104 | |||
| XM_015430669.1 | XP_015286155.1 | 7 - 8 | 104 | |||
| GenBank mRNA | AB169620.1 | - | 9 - 10 | 104 | ||
| AB171299.1 | - | 9 - 10 | 901 | 104 | ||
More Information
Set Membership: |
|

NAP1L1
Genomic Map