Assay Details
| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284057.1 | NP_001270986.1 | 3 - 4 | 494 | 76 | |
| XM_015440919.1 | XP_015296405.1 | 6 - 7 | 1527 | 77 | ||
| XM_015440920.1 | XP_015296406.1 | 5 - 6 | 1098 | 77 | ||
| XM_015440921.1 | XP_015296407.1 | 6 - 7 | 1524 | 77 | ||
| XM_015440922.1 | XP_015296408.1 | 6 - 7 | 1043 | 77 | ||
| XM_015440923.1 | XP_015296409.1 | 6 - 7 | 1046 | 77 | ||
| GenBank mRNA | AB171154.1 | - | 3 - 4 | 494 | 76 | |
More Information
Set Membership: |
|

KCTD15
Genomic Map