Assay Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284061.1 | NP_001270990.1 | 2 - 3 | 1072 | 65 | |
| GenBank mRNA | AB170829.1 | - | 2 - 3 | 1072 | 65 | |
More Information
Set Membership: |
|

CX3CR1
Genomic Map