Assay Details
Gene Symbol: LY6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285284.1 | NP_001272213.1 | 2 - 3 | 211 | 90 | |
XM_005564206.2 | XP_005564263.1 | 2 - 3 | 227 | 90 | ||
GenBank mRNA | AB169515.1 | - | 2 - 3 | 211 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
