Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285302.1 | NP_001272231.1 | 2 - 3 | 579 | 96 | |
| XM_005573923.2 | XP_005573980.1 | 2 - 3 | 968 | 96 | ||
| XM_015432056.1 | XP_015287542.1 | 2 - 3 | 969 | 96 | ||
| GenBank mRNA | AB179300.1 | - | 2 - 3 | 582 | 96 | |
More Information
Set Membership: |
|

FLACC1
Genomic Map