Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285317.1 | NP_001272246.1 | 12 - 13 | 2125 | 64 | |
| XM_005544665.2 | XP_005544722.1 | 12 - 13 | 2194 | 64 | ||
| XM_005544667.1 | XP_005544724.1 | 11 - 12 | 2086 | 64 | ||
| GenBank mRNA | AB071053.1 | - | 1482 | 64 | ||
| AB168993.1 | - | 12 - 13 | 2125 | 64 | ||
More Information
Set Membership: |
|

SPATA21
Genomic Map