Assay Details
| Gene Symbol: TRIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283153.1 | NP_001270082.1 | 5 - 6 | 704 | 63 | |
| XM_015451498.1 | XP_015306984.1 | 5 - 6 | 1146 | 63 | ||
| XM_015451499.1 | XP_015306985.1 | 5 - 6 | 717 | 63 | ||
| GenBank mRNA | AB170647.1 | - | 5 - 6 | 704 | 63 | |
More Information
Set Membership: |
|

TRIT1
Genomic Map