Assay Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283267.1 | NP_001270196.1 | 1 - 2 | 325 | 90 | |
| GenBank mRNA | AB173314.1 | - | 1 - 2 | 325 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLCXD3| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283267.1 | NP_001270196.1 | 1 - 2 | 325 | 90 | |
| GenBank mRNA | AB173314.1 | - | 1 - 2 | 325 | 90 | |