Assay Details
| Gene Symbol: PFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283582.1 | NP_001270511.1 | 1 - 2 | 254 | 104 | |
| GenBank mRNA | AB169846.1 | - | 1 - 2 | 254 | 104 | |
| AB174369.1 | - | 1 - 2 | 144 | 104 | ||
More Information
Set Membership: |
|

PFN2
Genomic Map