Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283871.1 | NP_001270800.1 | 11 - 12 | 3541 | 107 | |
| XM_015440941.1 | XP_015296427.1 | 11 - 12 | 3701 | 107 | ||
| XM_015440942.1 | XP_015296428.1 | 12 - 13 | 5169 | 107 | ||
| XM_015440943.1 | XP_015296429.1 | 12 - 13 | 3773 | 107 | ||
| XM_015440944.1 | XP_015296430.1 | 14 - 15 | 5108 | 107 | ||
| XM_015440945.1 | XP_015296431.1 | 11 - 12 | 5106 | 107 | ||
| XM_015440946.1 | XP_015296432.1 | 12 - 13 | 3095 | 107 | ||
| GenBank mRNA | AB179365.1 | - | 11 - 12 | 3541 | 107 | |
More Information
Set Membership: |
|

GARRE1
Genomic Map