Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283871.1 | NP_001270800.1 | 13 - 14 | 3758 | 67 | |
| XM_015440941.1 | XP_015296427.1 | 13 - 14 | 3918 | 67 | ||
| XM_015440942.1 | XP_015296428.1 | 14 - 15 | 5386 | 67 | ||
| XM_015440943.1 | XP_015296429.1 | 14 - 15 | 3990 | 67 | ||
| XM_015440944.1 | XP_015296430.1 | 16 - 17 | 5325 | 67 | ||
| XM_015440945.1 | XP_015296431.1 | 13 - 14 | 5323 | 67 | ||
| XM_015440946.1 | XP_015296432.1 | 14 - 15 | 3312 | 67 | ||
| GenBank mRNA | AB179365.1 | - | 13 - 14 | 3758 | 67 | |
More Information
Set Membership: |
|

GARRE1
Genomic Map