Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283871.1 | NP_001270800.1 | 2 - 3 | 1345 | 81 | |
| XM_015440941.1 | XP_015296427.1 | 2 - 3 | 1505 | 81 | ||
| XM_015440942.1 | XP_015296428.1 | 3 - 4 | 2973 | 81 | ||
| XM_015440943.1 | XP_015296429.1 | 3 - 4 | 1577 | 81 | ||
| XM_015440944.1 | XP_015296430.1 | 5 - 6 | 2912 | 81 | ||
| XM_015440945.1 | XP_015296431.1 | 2 - 3 | 2910 | 81 | ||
| XM_015440946.1 | XP_015296432.1 | 3 - 4 | 899 | 81 | ||
| GenBank mRNA | AB179365.1 | - | 2 - 3 | 1345 | 81 | |

GARRE1
Genomic Map