Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283871.1 | NP_001270800.1 | 3 - 4 | 1558 | 87 | |
| XM_015440941.1 | XP_015296427.1 | 3 - 4 | 1718 | 87 | ||
| XM_015440942.1 | XP_015296428.1 | 4 - 5 | 3186 | 87 | ||
| XM_015440943.1 | XP_015296429.1 | 4 - 5 | 1790 | 87 | ||
| XM_015440944.1 | XP_015296430.1 | 6 - 7 | 3125 | 87 | ||
| XM_015440945.1 | XP_015296431.1 | 3 - 4 | 3123 | 87 | ||
| XM_015440946.1 | XP_015296432.1 | 4 - 5 | 1112 | 87 | ||
| GenBank mRNA | AB179365.1 | - | 3 - 4 | 1558 | 87 | |

GARRE1
Genomic Map