Assay Details
| Gene Symbol: GARRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283871.1 | NP_001270800.1 | 6 - 7 | 1956 | 64 | |
| XM_015440941.1 | XP_015296427.1 | 6 - 7 | 2116 | 64 | ||
| XM_015440942.1 | XP_015296428.1 | 7 - 8 | 3584 | 64 | ||
| XM_015440943.1 | XP_015296429.1 | 7 - 8 | 2188 | 64 | ||
| XM_015440944.1 | XP_015296430.1 | 9 - 10 | 3523 | 64 | ||
| XM_015440945.1 | XP_015296431.1 | 6 - 7 | 3521 | 64 | ||
| XM_015440946.1 | XP_015296432.1 | 7 - 8 | 1510 | 64 | ||
| GenBank mRNA | AB179365.1 | - | 6 - 7 | 1956 | 64 | |
More Information
Set Membership: |
|

GARRE1
Genomic Map