Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283982.1 | NP_001270911.1 | 2 - 3 | 156 | 68 | |
| XM_005592640.2 | XP_005592697.1 | 2 - 3 | 395 | 68 | ||
| XM_005592641.2 | XP_005592698.1 | 2 - 3 | 183 | 68 | ||
| XM_005592642.2 | XP_005592699.1 | 2 - 3 | 1699 | 68 | ||
| XM_005592643.2 | XP_005592700.1 | 4 - 5 | 5436 | 68 | ||
| GenBank mRNA | AB168754.1 | - | 2 - 3 | 156 | 68 | |
More Information
Set Membership: |
|

SDR42E1
Genomic Map