Assay Details
| Gene Symbol: MAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284558.1 | NP_001271487.1 | 3 - 4 | 473 | 69 | |
| GenBank mRNA | AB171631.1 | - | 3 - 4 | 473 | 69 | |
More Information
Set Membership: |
|

MAL
Genomic Map