Assay Details
| Gene Symbol: NSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285058.1 | NP_001271987.1 | 3 - 4 | 437 | 71 | |
| GenBank mRNA | AB174648.1 | - | 3 - 4 | 437 | 71 | |
More Information
Set Membership: |
|

NSG1
Genomic Map