Assay Details
| Gene Symbol: BPGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285135.1 | NP_001272064.1 | 3 - 4 | 871 | 106 | |
| XM_005550827.2 | XP_005550884.1 | 2 - 3 | 818 | 106 | ||
| GenBank mRNA | AB169166.1 | - | 3 - 4 | 871 | 106 | |
More Information
Set Membership: |
|

BPGM
Genomic Map