Assay Details
| Gene Symbol: NUP50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285194.1 | NP_001272123.1 | 8 - 9 | 1461 | 64 | |
| XM_015456944.1 | XP_015312430.1 | 7 - 8 | 1532 | 64 | ||
| XM_015456945.1 | XP_015312431.1 | 7 - 8 | 1339 | 64 | ||
| GenBank mRNA | AB168260.1 | - | 8 - 9 | 1461 | 64 | |
| AB179101.1 | - | 7 - 8 | 1246 | 64 | ||
More Information
Set Membership: |
|

NUP50
Genomic Map