Assay Details
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283374.1 | NP_001270303.1 | 3 - 4 | 527 | 67 | |
| XM_015450074.1 | XP_015305560.1 | 3 - 4 | 674 | 67 | ||
| GenBank mRNA | AB173891.1 | - | 3 - 4 | 527 | 67 | |
| HM769337.1 | - | 3 - 4 | 447 | 67 | ||
More Information
Set Membership: |
|

BST1
Genomic Map