Assay Details
| Gene Symbol: CABYR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283766.1 | NP_001270695.1 | 4 - 5 | 1964 | 79 | |
| XM_015439789.1 | XP_015295275.1 | 5 - 6 | 2258 | 79 | ||
| XM_015439790.1 | XP_015295276.1 | 6 - 7 | 2338 | 79 | ||
| XM_015439791.1 | XP_015295277.1 | 5 - 6 | 2318 | 79 | ||
| XM_015439792.1 | XP_015295278.1 | 5 - 6 | 1784 | 79 | ||
| XM_015439793.1 | XP_015295279.1 | 4 - 5 | 2181 | 79 | ||
| XM_015439794.1 | XP_015295280.1 | 5 - 6 | 1324 | 79 | ||
| XM_015439795.1 | XP_015295281.1 | 6 - 7 | 1383 | 79 | ||
| XM_015439796.1 | XP_015295282.1 | 4 - 5 | 1242 | 79 | ||
| XM_015439797.1 | XP_015295283.1 | 5 - 6 | 845 | 79 | ||
| GenBank mRNA | AB179126.1 | - | 2 - 3 | 1834 | 79 | |
| AB179138.1 | - | 5 - 6 | 1696 | 79 | ||
| AB179372.1 | - | 4 - 5 | 1964 | 79 | ||
More Information
Set Membership: |
|

CABYR
Genomic Map