Assay Details
| Gene Symbol: GOLT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284693.1 | NP_001271622.1 | 4 - 5 | 480 | 79 | |
| GenBank mRNA | AB174429.1 | - | 4 - 5 | 480 | 79 | |
More Information
Set Membership: |
|

GOLT1B
Genomic Map