Assay Details
| Gene Symbol: RAB11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285131.1 | NP_001272060.1 | 3 - 4 | 498 | 94 | |
| XM_015440147.1 | XP_015295633.1 | 3 - 4 | 647 | 93 | ||
| GenBank mRNA | AB169492.1 | - | 3 - 4 | 498 | 94 | |
| AB174302.1 | - | 3 - 4 | 486 | 93 | ||
More Information
Set Membership: |
|

RAB11B
Genomic Map