Assay Details
| Gene Symbol: LMNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285366.1 | NP_001272295.1 | 4 - 5 | 764 | 119 | |
| XM_015430157.1 | XP_015285643.1 | 4 - 5 | 771 | 119 | ||
| XM_015430158.1 | XP_015285644.1 | 4 - 5 | 753 | 119 | ||
| XM_015430159.1 | XP_015285645.1 | 4 - 5 | 629 | 119 | ||
| XM_015430160.1 | XP_015285646.1 | 4 - 5 | 554 | 119 | ||
| GenBank mRNA | AB168558.1 | - | 4 - 5 | 764 | 119 | |
More Information
Set Membership: |
|

LMNTD1
Genomic Map