Assay Details
| Gene Symbol: LMNTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285366.1 | NP_001272295.1 | 6 - 7 | 1070 | 96 | |
| XM_015430157.1 | XP_015285643.1 | 6 - 7 | 1077 | 96 | ||
| XM_015430158.1 | XP_015285644.1 | 6 - 7 | 1059 | 96 | ||
| XM_015430159.1 | XP_015285645.1 | 6 - 7 | 935 | 96 | ||
| XM_015430160.1 | XP_015285646.1 | 6 - 7 | 860 | 96 | ||
| GenBank mRNA | AB168558.1 | - | 6 - 7 | 1070 | 96 | |
More Information
Set Membership: |
|

LMNTD1
Genomic Map