Assay Details
| Gene Symbol: POLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283565.1 | NP_001270494.1 | 6 - 7 | 1280 | 62 | |
| XM_005549689.2 | XP_005549746.1 | 6 - 7 | 931 | 62 | ||
| XM_005549690.2 | XP_005549747.1 | 6 - 7 | 1146 | 62 | ||
| XM_015447310.1 | XP_015302796.1 | 6 - 7 | 1454 | 62 | ||
| GenBank mRNA | AB169401.1 | - | 6 - 7 | 1280 | 62 | |
More Information
Set Membership: |
|

POLD2
Genomic Map