Assay Details
| Gene Symbol: GLIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283670.1 | NP_001270599.1 | 5 - 6 | 456 | 72 | |
| XM_005581329.2 | XP_005581386.1 | 4 - 5 | 602 | 72 | ||
| GenBank mRNA | AB168400.1 | - | 5 - 6 | 456 | 72 | |
More Information
Set Membership: |
|

GLIPR2
Genomic Map