Assay Details
| Gene Symbol: SPAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283703.1 | NP_001270632.1 | 3 - 4 | 1358 | 95 | |
| XM_005550650.2 | XP_005550707.1 | 4 - 5 | 1470 | 95 | ||
| XM_015447311.1 | XP_015302797.1 | 5 - 6 | 1586 | 95 | ||
| GenBank mRNA | AB169283.1 | - | 3 - 4 | 1359 | 95 | |
| L13780.1 | - | 1 - 2 | 963 | 95 | ||
More Information
Set Membership: |
|

SPAM1
Genomic Map