Assay Details
| Gene Symbol: GDAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283862.1 | NP_001270791.1 | 7 - 8 | 1080 | 85 | |
| GenBank mRNA | AB169311.1 | - | 7 - 8 | 1080 | 85 | |
More Information
Set Membership: |
|

GDAP2
Genomic Map