Assay Details
| Gene Symbol: DNASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284852.1 | NP_001271781.1 | 5 - 6 | 834 | 58 | |
| XM_015440152.1 | XP_015295638.1 | 5 - 6 | 922 | 58 | ||
| GenBank mRNA | AB170205.1 | - | 5 - 6 | 727 | 58 | |
| AB170800.1 | - | 5 - 6 | 834 | 58 | ||
| AB170812.1 | - | 5 - 6 | 812 | 58 | ||
More Information
Set Membership: |
|

DNASE2
Genomic Map