Assay Details
| Gene Symbol: BDKRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284883.1 | NP_001271812.1 | 1 | 1015 | 71 | |
| XM_015453260.1 | XP_015308746.1 | 2 | 2392 | 71 | ||
| GenBank mRNA | AY788905.1 | - | 1 | 1015 | 71 | |
More Information
Set Membership: |
|

BDKRB1
Genomic Map