Assay Details
| Gene Symbol: ATP5PF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284898.1 | NP_001271827.1 | 1 - 2 | 162 | 121 | |
| XM_015446615.1 | XP_015302101.1 | 2 - 3 | 268 | 121 | ||
| XM_015446616.1 | XP_015302102.1 | 2 - 3 | 341 | 121 | ||
| XM_015446617.1 | XP_015302103.1 | 2 - 3 | 283 | 121 | ||
| GenBank mRNA | AB072025.1 | - | 1 - 2 | 162 | 121 | |
More Information
Set Membership: |
|

ATP5PF
Genomic Map