Assay Details
| Gene Symbol: MTNR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285066.1 | NP_001271995.1 | 1 - 2 | 226 | 58 | |
| XM_015435347.1 | XP_015290833.1 | 1 - 2 | 323 | 58 | ||
| GenBank mRNA | FJ905899.1 | - | 1 - 2 | 226 | 58 | |
More Information
Set Membership: |
|

MTNR1B
Genomic Map