Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285128.1 | NP_001272057.1 | 1 - 2 | 255 | 103 | |
| GenBank mRNA | AB174639.1 | - | 1 - 2 | 255 | 103 | |
More Information
Set Membership: |
|

C1H1orf43
Genomic Map