Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285128.1 | NP_001272057.1 | 4 - 5 | 532 | 57 | |
| XM_005541675.2 | XP_005541732.1 | 3 - 4 | 626 | 57 | ||
| XM_015452008.1 | XP_015307494.1 | 4 - 5 | 651 | 57 | ||
| GenBank mRNA | AB174639.1 | - | 4 - 5 | 532 | 57 | |
More Information
Set Membership: |
|

C1H1orf43
Genomic Map