Assay Details
| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285229.1 | NP_001272158.1 | 2 - 3 | 330 | 54 | |
| XM_015454365.1 | XP_015309851.1 | 2 - 3 | 739 | 54 | ||
| XM_015454366.1 | XP_015309852.1 | 2 - 3 | 737 | 54 | ||
| XM_015454367.1 | XP_015309853.1 | 2 - 3 | 736 | 54 | ||
| GenBank mRNA | AB171011.1 | - | 2 - 3 | 330 | 54 | |
More Information
Set Membership: |
|

DUSP26
Genomic Map