Assay Details
| Gene Symbol: ASAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285344.1 | NP_001272273.1 | 4 - 5 | 352 | 78 | |
| GenBank mRNA | AB125153.1 | - | 4 - 5 | 366 | 78 | |
| AB174322.1 | - | 4 - 5 | 352 | 78 | ||
More Information
Set Membership: |
|

ASAH1
Genomic Map