Assay Details
| Gene Symbol: SQLE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287728.1 | NP_001274657.1 | 2 - 3 | 706 | 76 | |
| GenBank mRNA | AB220561.1 | - | 2 - 3 | 706 | 76 | |
More Information
Set Membership: |
|

SQLE
Genomic Map