Assay Details
| Gene Symbol: CYP2C19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283290.1 | NP_001270219.1 | 7 - 8 | 1177 | 118 | |
| XM_015455918.1 | XP_015311404.1 | 7 - 8 | 118 | |||
| GenBank mRNA | DQ074805.1 | - | 7 - 8 | 1177 | 118 | |
More Information
Set Membership: |
|

CYP2C19
Genomic Map