Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284721.1 | NP_001271650.1 | 3 - 4 | 350 | 121 | |
| GenBank mRNA | AB173089.1 | - | 3 - 4 | 350 | 121 | |
More Information
Set Membership: |
|

NUTF2
Genomic Map