Assay Details
| Gene Symbol: MRPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283946.1 | NP_001270875.1 | 4 | 519 | 130 | |
| XM_005583667.2 | XP_005583724.1 | 4 | 718 | 130 | ||
| GenBank mRNA | AB173629.1 | - | 4 | 519 | 130 | |
More Information
Set Membership: |
|

MRPL27
Genomic Map